Would you like to log in again?
Clorox® 4 in One Disinfectant and Sanitizer, Lavender, Essendant
Supplier: Essendant
Clorox®
CLO32512
76500-154CT 106.85 USD InStock
76500-154
Clorox® 4 in One Disinfectant and Sanitizer, Lavender, Essendant
Cleaning Agents Disinfectants
Clorox 4 in One Disinfectant and Sanitizer disinfects, sanitizes, and reduces allergens with a single all-purpose aerosol spray.
  • EPA-registered to kill more than 20 bacteria, virus, and fungi in five minutes on hard surfaces
  • EPA-registered to kill bacteria on soft surfaces including rugs, mattresses, curtains, couches, and more
  • Create a healthier environment by sanitizing the air and killing odor-causing bacteria, leaving behind a refreshing scent
  • Efficiently reduces many common causes of allergies, from dog and cat dander to dust mites and cockroach matter
  • Chemical compound: Ethanol 66.34%; Dipropylene Glycol 5.31%
This powerful cleaner disinfects hard surfaces with 24 EPA-registered kill claims, and sanitizes soft surfaces in as little as 30 seconds. It meets EPA criteria for use against SARS-CoV-2, the virus that causes COVID-19. This disinfecting spray even kills odor-causing bacteria in the air, and leaves behind a refreshing scent. On top of that, Clorox 4 in One reduces pet dander, dust mite matter and cockroach matter allergens which can trigger allergies and asthma.
Ideal for use in offices, day care centers, schools, hotels, restaurants, and other commercial facilities. No precleaning required.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty