Would you like to log in again?
BioClean™ Non Sterile DB Pouch-Style Face Mask, Ansell
Supplier: Ansell
BioClean™
BDBN
76521-426CS 171.56 USD InStock
76521-426
BioClean™ Non Sterile DB Pouch-Style Face Mask, Ansell
Masks Cleanroom Masks
Non sterile pouch-style face mask.
  • High bacterial and particle filtration efficiency
  • Large breathing chamber
  • Ultrasonically sealed edges
  • Fully enclosed malleable nose-band
  • Inner and outer facing layer: non-woven spunbonded polyester (hygroscopic)
The BioClean™ DB Pouch-style face mask features a large breathing chamber for increased wearer comfort and a malleable noseband for a secure fit. Made from cleanroom compatible materials and ultrasonically sealed edges to reduce the risk of contamination entering the controlled environment.
Certifications: ISO 2859, ISO 9001:2015.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty