Would you like to log in again?
Buccutite™ Peroxidase (HRP) Antibody Conjugation Kits (2 Labelings), AAT Bioquest
Supplier: AAT Bioquest
5504 5505 5503 5506
76481-790EA 288.22 USD InStock
76481-790 76484-792 76484-464 76484-794
Buccutite™ Peroxidase (HRP) Antibody Conjugation Kits (2 Labelings), AAT Bioquest
Protein Linking and Labeling Systems
Protein-protein conjugations are commonly performed with a bifunctional linker (such as the commonly used SMCC), having different reactivity on each end for linking two different proteins.
  • High conjugation yield
  • Stable crosslinking
  • Minimal hands-on time
  • Robust protocol
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty