Synonyms:
O6-(4-Aminomethyl-benzyl)guanine
12560
12561
76485-232EA
483.08
USD
InStock
76485-232
76485-230
BG-NH2 for SNAP-tag fusion
BG-NH2
BG-NH2 is a carboxy-reactive building block for preparing of SNAP substrates from the NHS esters or other amine-reactive forms of labels or biological targets.
- SNAP substrate building block
- Carboxy-reactive