Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Digoxigenin-11-dUTP 1 mM for PCR

Supplier: AAT BIOQUEST INC
Warning

17012
76484-600EA 180.84 USD InStock
76484-600
Digoxigenin-11-dUTP 1 mM for PCR
Digoxigenin-11-dUTP
Digoxigenin-11-dUTP can be enzymatically incorporated into DNA/cDNA as substitute for its natural counterpart dTTP.

  • Limited DIG interference
  • High quality


MW: 1132,87 g/mol
CAS Number: 129030-36-0
ADR: 5,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/32/24/43933224.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty