Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

iFluor® 488, Alkyne fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

999
76484-546EA 240.68 USD InStock
76484-546
iFluor® 488, Alkyne fluorescent probe
iFluor®
Although FITC is still the most popular fluorescent labeling dye for preparing green fluorescent bioconjugates, there are certain limitations with FITC, such as severe photobleaching for microscope imaging and pH-sensitive fluorescence.

  • pH-insensitive
  • Azide reactive
  • High photostability
  • Brighter signal


ADR: 5,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/32/22/43933222.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty