Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

iFluor® 488 fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

1005
76485-602EA 229.01 USD InStock
76485-602
iFluor® 488 fluorescent probe
iFluor®
The tetrazine-trans-cyclooctene (TCO) ligation constitutes a non-toxic biomolecule labeling method of unparalleled speed.

  • pH-insensitive
  • Azide reactive
  • High photostability
  • Brighter signal


ADR: 5,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/33/13/43933313.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty