Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Green 630 succinimidyl ester, mFluor™ fluorescent probe

Supplier: AAT BIOQUEST INC
Warning
Synonyms: Green 630 SE

1168
76485-578EA 344.92 USD InStock
76485-578
Green 630 succinimidyl ester, mFluor™ fluorescent probe
Green 630 succinimidyl ester
Advances in spectral flow cytometers have expanded applications and capabilities beyond conventional flow cytometry.

  • pH-insensitive
  • High photostability
  • Brighter signal


MW: 806.94 g/mol
ADR: 5,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/33/04/43933304.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty