Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

XFD350 NHS Ester fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

1800
76484-518EA 240.68 USD InStock
76484-518
XFD350 NHS Ester fluorescent probe
XFD350 NHS Ester
XFD350 NHS Ester (Succinimidyl Ester) is the same molecule to the Alexa Fluor® 350 NHS Ester (Alexa Fluor® is the trademark of ThermoFisher).

  • pH-insensitive
  • High photostability
  • Brighter signal


MW: 410.35 g/mol
ADR: 5,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty