Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Biotin-PEG2-NHS biodetection

Supplier: AAT BIOQUEST INC
Warning
Synonyms: 2,5-Dioxopyrrolidin-1-yl 3-(2-(2-(5-(2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamido)ethoxy)ethoxy)propanoateBiotin-PEG2-NHS ester9-(Biotinamido)-4,7-dioxanonanoic acid N-Succinimidyl esterN-Succinimidyl 9-(Biotinamido)-4,7-dioxanonanoate

3016
76481-626EA 118.54 USD InStock
76481-626
Biotin-PEG2-NHS biodetection
Biotin-PEG2-NHS
This amine-reactive biotin derivative contains a long arm (~20 Å) to increase its avidin-binding affinity.

  • Improved labeling efficiency
  • High binding affinity


Formula: C₂₁H₃₂N₄O₈S
MW: 500.57 g/mol
MDL Number: MFCD00467154
CAS Number: 596820-83-6
ADR: 5,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/30/66/43933066.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty