Would you like to log in again?
FMOC-Lys(TF3)-OH for biomolecular labeling
Supplier: AAT BIOQUEST INC
Warning
5051
76481-742EA 604.2 USD InStock
76481-742
FMOC-Lys(TF3)-OH for biomolecular labeling
FMOC-Lys(TF3)-OH
FMOC-Lys(TF3)-OH is a building block for in-sequence Lys labeling by TF3.
  • FRET building block
  • Multiple applications
ADR: 5,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/31/03/43933103.jpg
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty