Would you like to log in again?
Fluorescein aldehyde [5-FAM aldehyde] for biomolecular labeling
Supplier: AAT BIOQUEST INC
Warning
9000
76485-704EA 344.92 USD InStock
76485-704
Fluorescein aldehyde [5-FAM aldehyde] for biomolecular labeling
Fluorescein aldehyde [5-FAM aldehyde]
Fluorescein aldehyde is a reactive fluorescent dye that can react with an amine, hydrazine or hydroxylamine.
  • Reactive with amine, hydrazine, or hydroxylamine
  • Detectable with the FITC channel
ADR: 5,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/33/25/43933325.jpg
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty