Would you like to log in again?
ReadiLink™ Rapid XFD555 Antibody Labeling Kit *XFD555 Same Structure to Alexa Fluor™ 555* (2×50 µg Labelings), AAT Bioquest
Supplier: AAT Bioquest
1276
76485-562EA 170.98 USD InStock
76485-562
ReadiLink™ Rapid XFD555 Antibody Labeling Kit *XFD555 Same Structure to Alexa Fluor™ 555* (2×50 µg Labelings), AAT Bioquest
Protein Linking and Labeling Systems
XFD555 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 555 (Alexa Fluor® is the trademark of ThermoFisher).
  • Contains all essential components
  • Minimal hands on time
  • Ready to use
  • Robust and simple antibody labeling protocol
  • No column purification required
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty