Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Violet 450-VAD-FMK, mFluor™
Supplier: AAT BIOQUEST INC
Warning
13475
76482-156EA 118.54 USD InStock
76482-156
Violet 450-VAD-FMK, mFluor™
Violet 450-VAD-FMK
FAM-VAD is a blue fluorescent cell-permeable polycaspase inhibitor to target caspases 1, 2, 3, 6, 8, 9, or 10.
  • caspase inhibitor
  • bright signal
  • multiple applications
ADR: 5,II
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty