Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tide Quencher™ 4WS maleimide, Tide Quencher™ fluorescent probe

Supplier: AAT BIOQUEST INC
Warning
Synonyms: TQ4WS maleimide

2064
76481-334EA 361.89 USD InStock
76481-334
Tide Quencher™ 4WS maleimide, Tide Quencher™ fluorescent probe
Tide Quencher™ 4WS maleimide
TQ4WS is designed to be a superior quencher to ROX, TF4 and Texas Red®.

  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling


MW: 1106,2 g/mol
ADR: 5,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/29/61/43932961.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty