Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tide Fluor™ 7WS amine, Tide Fluor™ fluorescent probe

Supplier: AAT BIOQUEST INC
Warning
Synonyms: TF7WS amine

2331
76481-524EA 240.68 USD InStock
76481-524
Tide Fluor™ 7WS amine, Tide Fluor™ fluorescent probe
Tide Fluor™ 7WS amine
Tide Fluor™ 7WS (TF7WS) family has the spectral properties similar to those of Cy7, IRDye 800 and Alexa Fluor 750.

  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity


MW: 1047.07 g/mol
ADR: 5,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty