Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Washable Glue, 16 oz.

Supplier: HANDY ART, INC. MO
211-190
470227-584EA 5.85 USD InStock
470227-584
Washable Glue, 16 oz.
Adhesives
All Handy Art glue is proudly made in the USA.

  • Bonds to most porous and non-porous materials
  • Non-toxic
  • Size: 16 oz. (473 ml)

Use as adhesive on cards, masks, wood crafts, scrapbooking, fabric, glass, felt, and more.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty