Would you like to log in again?
GP Forward SC General-Purpose Cleaner, Diversey
Supplier: Diversey
GP Forward SC
904965
76514-346CS 185.06 USD InStock
76514-346
GP Forward SC General-Purpose Cleaner, Diversey
Cleaning Agents Detergents and Surface Cleansers
Concentrated formulations deliver powerful cleaning performance for just pennies per diluted gallon.
  • A high-performance cleaner for daily use on floors, walls and all washable surfaces
  • Removes all types of dirt, grease and soap scum with no rinsing required
  • Works with cold, hot and soft/hard water in mop, bucket, autoscrubber and trigger spray applications
  • Green in color with a pleasant citrus scent
A highly versatile general cleaner concentrate that contains a blend of high-performance detergents to effectively clean surfaces. Can be used on any water-safe surface, and has a pleasant citrus scent. Excellent for use in scrub and recoat floor care programs. Green in color with a citrus scent.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty