Would you like to log in again?
Stride Floral HC Neutral Cleaner, Diversey
Supplier: Diversey
Stride Floral HC
904717
76514-362CS 458.12 USD InStock
76514-362
Stride Floral HC Neutral Cleaner, Diversey
Cleaning Agents Detergents and Surface Cleansers
Concentrated formulation delivers economical cleaning performance.
  • Low-foaming formula allows for less emptying of recovery tanks and increased productivity
  • Neutral pH formulation does not require rinsing and dries film-free
  • For use as a daily and general cleaner
  • Versatile formula that performs in a variety of application methods such as an autoscrubber, spray, wipe and mop and bucket
  • Green Seal certification
A neutral cleaner concentrate formulated for everyday cleaning of floors and other hard surfaces. This product does not dull floor finishes and does not require rinsing to deliver labor savings. Leaves a pleasant floral fragrance in cleaned areas. Pink in color with a floral scent.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty