Would you like to log in again?
Viral Transport Medium (VTM), Hardy Diagnostics
Supplier: Hardy Diagnostics
R64BX
76523-368PK 236.46 USD InStock
76523-368
Viral Transport Medium (VTM), Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
Hardy Diagnostics viral transport medium is recommended for the collection and transport of clinical specimens for the recovery of viral agents.
  • Hardy Diagnostics VTM does not contain guanidium thiocyanate
  • Contains a pH indicator
  • Selective agents are added to inhibit bacterial and fungal contaminants
  • Medium acts as a cryoprotectant to ensure virus stabilization through freezing and thawing
Hardy Diagnostics viral transport medium is recommended for the collection and transport of clinical specimens for the recovery of viral agents including, but not limited to, Herpes Simplex Type I, Herpes Simplex Type II, Cytomegalovirus (CMV), Influenza A, Influenza B, Respiratory Syncytial Virus (RSV), Echovirus, Adenovirus, etc.
Media can be stored at room temperature (RT) 2-30 °C  without decreased performance. Data permitting, the length of RT storage shall continue to be extended as the real-time findings are collected, and applied to each lot. For the length of allowable RT storage (post retrieval from 2-30 °C conditions), please refer to the respective Certificate of Analysis for the lot-specific information.
Ordering information: Swabs not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty