Would you like to log in again?
Vikan® High-Temp Detail Brush, Remco
Supplier: Remco
Vikan®
44023
76528-986EA 10.6 USD InStock
76528-986
Vikan® High-Temp Detail Brush, Remco
Brushes
This brush can remove debris from heat sealers and other packing equipment.
  • Extra stiff bristles
  • Polypropylene block, polyester bristles
High temperature resistance also allows cleaning small gaps in grills or other equipment while they are still warm.
Overall dimensions: 8.07L×0.79W×2.36"H
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty