Would you like to log in again?
Vikan® Tank Brushes, Stiff Bristles, Remco
Supplier: Remco
Vikan®
703766 703777 703788
76530-306EA 39.43 USD InStock
76530-306 76530-304 76530-308
Vikan® Tank Brushes, Stiff Bristles, Remco
Brushes
This oval-shaped brush with stiff bristles is effective for scrubbing inside large industrial tanks and other large container-like equipment.
  • Polyester bristles, polypropylene block
  • Euro (hygienic) thread type
The rounded edges work well to clean down the sides of equipment. It also fits well into large channel drains.
Overall dimensions: 8.3L× 5.1W×3.7"H
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty