Would you like to log in again?
United Scientific Supplies Dropper Bottle, Unassembled, LDPE, 15 ml
Supplier: United Scientific Supplies
PDB015-U-PK/24
470332-692PK 9.4 USD InStock
470332-692
United Scientific Supplies Dropper Bottle, Unassembled, LDPE, 15 ml
Bottles Dropping and Dispensing Bottles
This dropper bottle is made of translucent and low density polyethylene.
  • Bottle can be used with or without dropper
  • Comes in a pack of 24
For use when dispensing small drops of a solution. The drop sizes are uniform for consistent dispensing of liquids.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty