Would you like to log in again?
VWR® Blood Bank Thermometer
Supplier: Avantor
VWR®
60805-0011
76491-642CS 388.37 USD InStock
76491-642
VWR® Blood Bank Thermometer
Thermometers Analog Thermometers
This thermometer has been designed to indicate critical temperature range.
  • Total immersion
  • Individual serial numbers
  • PTFE coating helps prevent liquid and glass contamination
The critical temperature range from 4 to 6 °C is marked with arrows.
Certifications: Traceable NIST. Envirokleen certified by ChemTel (an independent laboratory).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty