Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Thermometer Clamp with Boss Head

Supplier: EISCO SCIENTIFIC LLC
CH0667A
470337-254EA 5.4 USD InStock
470337-254
Thermometer Clamp with Boss Head
Clamps General Purpose Clamps
This thermometer clamp has adjustable 360° jaw rotation.

  • Accommodates thermometers 6 to 12 mm in diameter
  • Overall length 140 mm

For attaching to support rods up to 16 mm in diameter. The clamp is die casted with thumbscrews and the jaws are made of phospher-bronze.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty