Would you like to log in again?
CloroxPro™ Disinfecting Wipes, Fresh Scent, Essendant
Supplier: Essendant
CLO31428 CLO01593CT
76500-234CT 92.95 USD InStock
76500-234 76500-112
CloroxPro™ Disinfecting Wipes, Fresh Scent, Essendant
Wipes Disposable Wipes and Towels Disinfectant Wipes
CloroxPro™ Disinfecting Wipes clean and disinfect in one easy step. These disinfecting wipes are EPA-registered to kill 99.9% of germs on hard non-porous surfaces, including cold and flu viruses, the COVID-19 virus (SARS-CoV-2), E. coli, Salmonella, Staph, Strep, MRSA and the organism that can cause whooping cough.
  • Disinfecting wipes are triple-layered to clean, disinfect, and remove allergens for 5x cleaning power.
  • Safe for finished wood, sealed granite, and stainless steel, and on non-food-contact surfaces
  • Disinfect and deodorize with an all-in-one cleaning alternative that’s safe to use on toys and more
  • Dissolves grease and removes tough soap scum
These wipes also remove common allergens like pet dander, pollen particles and grass. One wipe kills most viruses in as little as 15 seconds on a wide variety of surfaces. The premoistened, thick, antibacterial wipes are perfect for spills and dirt on a variety of hard, non-porous surfaces including acrylic, glass, and vinyl. They quickly clean and disinfect high-touch germ hot spots such as doorknobs, handrails, and faucets and are suitable for use on phones and keyboards. These sanitizing wipes are ideal for use in shared public spaces, offices, athletic facilities, schools, day care centers, busy healthcare environments, long term care, and other commercial facilities.
Chemical compound: Hexoxyethanol; Alkyl C12-18 Dimethylbenzyl Ammonium Chloride; Alkyl C12-14 Dimethyl Ethylbenzyl Ammonium Chloride; C12-14 Alcohols Ethoxylated Proproxylate; Isopropanol; Citric Acid.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty