Would you like to log in again?
Boardwalk® Microfiber Cleaning Kit, Essendant
Supplier: Essendant
Boardwalk®
BWKMFKIT
76500-300KT 77 USD InStock
76500-300
Boardwalk® Microfiber Cleaning Kit, Essendant
Mops
Boardwalk® Microfiber Cleaning Kit is a complete cleaning kit that includes everything needed to start a microfiber cleaning program.
  • Complete starter kit for microfiber cleaning system
  • Split microfibers allow for aggressive cleaning and removal of microscopic debris
  • Convenient telescopic handle extends from 35" to 60", clips easily onto frame
  • Colorflag coding for HACCP compliance
  • 18" mop heads
Split microfibers allow for more aggressive cleaning and removal of microscopic debris than conventional cleaning methods. The aluminum telescopic handle extends from 35" to 60" and clips easily onto the frame. Cloths and pads withstand hundreds of launderings, providing durability and strength.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty