Would you like to log in again?
Cole-Parmer® Essentials Wide-Mouth Plastic Bottles, PPCO, Antylia Scientific
Supplier: Antylia Scientific
06048-02 06048-20 06048-01 06048-10 06048-40 06048-30
76534-218PK 103.26 USD InStock
76534-218 76534-216 76534-214 76534-212 76534-220 76534-210
Cole-Parmer® Essentials Wide-Mouth Plastic Bottles, PPCO, Antylia Scientific
Bottles Plastic Bottles
Ideal for sensitive sample requirements.
  • Leakproof screw caps
  • Autoclavable
PP screw closures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty