Would you like to log in again?
Cole-Parmer® Essentials Graduated Square Plastic Bottles, PPCO, Antylia Scientific
Supplier: Antylia Scientific
06251-10 06251-50 06251-30 06251-20 06251-00 06251-40
76534-272PK 57.97 USD InStock
76534-272 76534-270 76534-268 76534-266 76534-276 76534-274
Cole-Parmer® Essentials Graduated Square Plastic Bottles, PPCO, Antylia Scientific
Bottles Plastic Bottles
Resistant to acids, alcohols, and bases.
  • Drip proof spout for easy pouring
  • Space-saving square design
  • Autoclavable
0 to 135 °C.
Ordering information: Caps sold separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty