Would you like to log in again?
PCRmax® Polypropylene PCR Microplates, Antylia Scientific
Supplier: Antylia Scientific
67103-53 67103-57 67103-54 67103-52
76533-890PK 427.79 USD InStock
76533-890 76533-892 76533-894 76533-896
PCRmax® Polypropylene PCR Microplates, Antylia Scientific
Microplates PCR Plates
Ultra-thin and consistent wall thickness allows precise thermal transfer for optimal results.
  • Manufactured with 99.9% pure virgin polypropylene
  • RNAse/DNAse-free, nonpyrogenic, non sterile
  • Compatible with sealing film
PCRmax® microplates are designed to fit real-time PCR thermal cyclers and sequencers. The construction of pure, clear polypropylene allows for full transparency. Microplates are compatible with PCRmax® Alpha Cycler instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty