Would you like to log in again?
Abdos® Cryo Boxes, 100 Place, Polycarbonate, Freezer Compatible, Foxx Life Sciences
Supplier: Foxx Life Sciences
Actero™ EZ
P20615A P20615 P20615W P20615B P20615Y
76521-928CS 72.22 USD InStock
76521-928 76521-926 76521-924 76521-932 76521-930
Abdos® Cryo Boxes, 100 Place, Polycarbonate, Freezer Compatible, Foxx Life Sciences
Boxes Cryoboxes
Manufactured from strong, autoclavable PC. These boxes create a small, space-efficient storage system while the specialized venting in the base and lid ensures a smooth flow for vapors and consistency in cooling.
  • Temperature range: –135...+135 °C
  • Transparent lids
  • Autoclavable: Yes
  • Shelf Life: 3 Years
The Cryo Box can be used in the gas phase of liquid nitrogen with appropriate safety procedures in place. The Transparent lid permit viewing of contents and has a numbered grid for sample identification. The dimensions of the Cryo Boxes are compatible with vertical, horizontal, and drawer-style freezer racks.
Certifications: Complies with: ISO 11137. Complies with: ISO Standard: 9001, 13485, 14001. Batch certificate included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty