Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Abdos® Serological Pipettes, Sterile, PS, Foxx Life Sciences

Supplier: Foxx Life Sciences
P10603 P10606 P10602 P10605 P10604 P10601
76521-958CS 213.35 USD InStock
76521-958 76521-968 76521-966 76521-964 76521-962 76521-960
Abdos® Serological Pipettes, Sterile, PS, Foxx Life Sciences
Pipettes Serological Pipettes
These serological pipettes have an advanced color-coded PU filter which offers complete protection against contamination.

  • Accuracy within ±2% at full volume
  • Color: Natural
  • Temperature range: +20...90 °C
  • Sterilization: E-beam
  • Material transparency: Clear
  • Shelf life: 3 years

The bidirectional graduations provide a choice of ascending and descending scales. The negative graduations allow additional working volume.

Certifications: Sterile Serological has been validated according to EN ISO 11137-2; 2012 for SAL 10-6. Complies with: ISO 11137. Complies with: ISO Standard: 9001, 13485, 14001. Batch certificate included.

Packaging: Individually wrapped with a paper/plastic peel-pack.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty