Would you like to log in again?
Axygen® 96-Well Tips, 200 µl, Clear, Filtered, Sterile, Wide Bore, SLAS Rack, Corning
Supplier: Corning
Axygen®
FXF-205-WB-R-S
76415-960CS 905 USD InStock
76415-960
Axygen® 96-Well Tips, 200 µl, Clear, Filtered, Sterile, Wide Bore, SLAS Rack, Corning
Pipette Tips
Axygen® offers a comprehensive line of tips for Beckman Coulter® automation systems. These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
  • 200 µl clear tips
  • 96 tip format
  • Clear polypropylene; wide bore
  • RNase-/DNase-free
  • Nonpyrogenic
  • With aerosol-resistant filters
  • Sterile
Clear polypropylene tips are available in 96-well and 384-well format designed for Beckman Coulter® Biomek® FX, Biomek® NX, Biomek® 3000 and Multimek™ AP-96, SPAN-8, MP20, MP200 and AP-384 head.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty