Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

ROS Brite™ DHCF

Supplier: AAT BIOQUEST INC
Warning

16053
76482-392EA 240.68 USD InStock
76482-392
ROS Brite™ DHCF
ROS Brite™
Reactive oxygen species (ROS) are chemically reactive molecules containing oxygen.

  • Bright fluorescence
  • Useful ros probe
  • Multiple research applications

Caution: For research use only


MW: 732,81 g/mol
ADR: 5,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty