Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DAX-J2™ IR

Supplier: AAT BIOQUEST INC
Warning

16302
76482-402EA 361.89 USD InStock
76482-402
DAX-J2™ IR
DAX-J2™
DAX-J2™ IR is a new nitric oxide (NO) sensor recently developed by AAT Bioquest.

  • Does not require esterase activity for no detection
  • Ph-independent fluorescence
  • More photostable than daf-2 product
  • More sensitive for detecting no than daf-2
  • Can be used for in vivo imaging of no

Caution: For research use only


MW: 476,57 g/mol
ADR: 5,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty