Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

iFluor® 488-dUTP 1 mM in TRIS buffer (pH 7.5)

Supplier: AAT BIOQUEST INC
Warning

17039
76485-814EA 107.38 USD InStock
76485-814
iFluor® 488-dUTP 1 mM in TRIS buffer (pH 7.5)
iFluor® 488-dUTP
The dye-modified deoxyuridine 5'-triphosphates are one of the most common methods to produce dye-labelled DNA via the conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labeling, or PCR.

  • Useful for DNA probe development
  • Well-cited
  • Multiple applications

Caution: For research use only


MW: 1152.83 g/mol
ADR: 5,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty