Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Propargylamino-3'-azidomethyl-dUTP

Supplier: AAT BIOQUEST INC
Warning

17093
76485-658EA 288.22 USD InStock
76485-658
5-Propargylamino-3'-azidomethyl-dUTP
5-Propargylamino-3'-azidomethyl-dUTP
5-Propargylamino-3'-azidomethyl-dUTP is a key building block for preparing fluorescent conjugates that are used in the next generation sequencing (NGS).

  • Ngs building block
  • Useful for multiple applications
  • High quality

Caution: For research use only


Formula: C13H19N6O14P3
MW: 576,24 g/mol
CAS Number:  666847-57-0
ADR: 5,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty