Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cal-520® maleimide

Supplier: AAT BIOQUEST INC
Warning

20610
76483-102EA 483.08 USD InStock
76483-102
Cal-520® maleimide
Cal-520® maleimide
Calcium measurement is critical for numerous biological investigations.

  • Calcium-dependent fluorescence
  • Steady signal
  • Improved photostability

Caution: For research use only


MW: 946.77 g/mol
ADR: 5,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty