Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

FURA-2 pentasodium salt

Supplier: AAT BIOQUEST INC
Warning

21026
76483-158EA 118.54 USD InStock
76483-158
FURA-2 pentasodium salt
FURA-2 pentasodium salt
Among the ratiometric calcium indicators, Fura-2 and Indo-1 are most commonly used.

  • Calcium-dependent fluorescence
  • Ratiometric detection
  • Cell impermeant

Caution: For research use only


Formula: C₂₉H₂₂N₃Na₅O₁₄
MW: 751.45 g/mol
MDL Number: MFCD00083328
CAS Number: 96314-98-6
ADR: 5,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty