Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rhod-5N tripotassium salt

Supplier: AAT BIOQUEST INC
Warning

21072
76483-216EA 300.63 USD InStock
76483-216
Rhod-5N tripotassium salt
Rhod-5N tripotassium salt
Calcium measurement is critical for numerous biological investigations.

  • Calcium-dependent fluorescence
  • Low binding affinity
  • Cell impermeant

Caution: For research use only


MW: 900.02 g/mol
ADR: 5,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty