Would you like to log in again?
Gelite™ Orange Nucleic Acid Gel Staining Kit, AAT Bioquest
Supplier: AAT Bioquest
17594
76482-932EA 240.68 USD InStock
76482-932
Gelite™ Orange Nucleic Acid Gel Staining Kit, AAT Bioquest
Assays Nucleic Acid Assays
Gelite™ Orange is an extremely sensitive nucleic acid gel stain for detecting DNA or RNA in gels using a standard 300 nm UV transilluminator and Polaroid 667 black-and-white print film.
  • High sensitivity
  • Exceptional DNA affinity
  • Robust protocol
  • Multiple applications
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty