Would you like to log in again?
Helixyte™ Green Fluorometric dsDNA Quantitation Kit *Optimized for Microplate Readers*, AAT Bioquest
Supplier: AAT Bioquest
17650
76482-944EA 240.68 USD InStock
76482-944
Helixyte™ Green Fluorometric dsDNA Quantitation Kit *Optimized for Microplate Readers*, AAT Bioquest
Assays Nucleic Acid Assays
Helixyte™ Green dsDNA Quantitation Assay Kit can be used for selectively detecting as little as 25 pg/ml of dsDNA in the presence of ssDNA, RNA, and free nucleotides.
  • High sensitivity
  • Exceptional DNA affinity
  • Robust protocol
  • Multiple applications
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty