Would you like to log in again?
MM 2-10 [N-(3-Triethylammoniumpropyl)-4-(4-(diethylamino)styryl)pyridinium dibromide] (1 mg), AAT Bioquest
Supplier: AAT Bioquest
21489
76483-402EA 118.54 USD InStock
76483-402
MM 2-10 [N-(3-Triethylammoniumpropyl)-4-(4-(diethylamino)styryl)pyridinium dibromide] (1 mg), AAT Bioquest
Stains Microbiology Stains
MM 2-10 (FM 2-10) is the abbreviation of our Membrane Marker 2-10, chemically called N-(3-triethylammoniumpropyl)-4-(4-(diethylamino)styryl)pyridinium dibromide.
  • Lipophilic vital stain
  • Adaptable and Robust
  • Multiple applications
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty