Would you like to log in again?
Di-2-ANEPEQ [JPW 1114] (5 mg), AAT Bioquest
Supplier: AAT Bioquest
21496
76483-408EA 240.68 USD InStock
76483-408
Di-2-ANEPEQ [JPW 1114] (5 mg), AAT Bioquest
Stains Microbiology Stains
Di-2-ANEPEQ is used for monitoring fast membrane potential changes.
  • Fast-response probe
  • Immediate optical response
  • Multiple applications
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty