Would you like to log in again?
CytoCalcein™ Violet 500 *Excited at 405 nm* (1 mg), AAT Bioquest
Supplier: AAT Bioquest
22013
76483-480EA 240.68 USD InStock
76483-480
CytoCalcein™ Violet 500 *Excited at 405 nm* (1 mg), AAT Bioquest
Stains Microbiology Stains
CytoCalcein™ Violet 500 is designed for labeling live cells in the same way to calcein, AM.
  • Optimized for flow cytometry excitation
  • Brighter signal
  • Suitable for Multiplexing
Caution: For research use only.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty