Would you like to log in again?
Ratchet Nut v3.0, MP Biomedicals
Supplier: MP Biomedicals
116005510
76522-172EA 231.04 USD InStock
76522-172
Ratchet Nut v3.0, MP Biomedicals
Wrenches
Accessory for FastPrep-24 and FastPrep-24 5G instruments. Securing Knob used to maintain the tube holder adapters on the central axis or 3 step assembly of the FastPrep-24 instruments.
  • FastPrep-24 5G: use with metal adapters
  • FastPrep-24 Classic: use with all adapters
Due to its high torque, the Ratchet Nut is particularly recommended for the heaviest Adapters, and especially for metal tube holders.
FastPrep-24 homogenizers can accommodate interchangeable adapters, which allow a unique versatility in sample number, size, and temperature conditions. All optional adapters stand stable on the benchtop and are commonly used as tube racks for sample storage at -20 or -80 °C.
Caution: Unless specified otherwise, MP Biomedicals' products are for research or further manufacturing use only, not for direct human use.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty