Would you like to log in again?
DiSBAC2(5) [Bis-(1,3-diethylthiobarbituric acid)pentamethine oxonol] (25 mg), AAT Bioquest
Supplier: AAT Bioquest
21415
76483-390EA 180.84 USD InStock
76483-390
DiSBAC2(5) [Bis-(1,3-diethylthiobarbituric acid)pentamethine oxonol] (25 mg), AAT Bioquest
Stains Microbiology Stains
DiSBAC2(5) is a sensitive slow-response membrane potential probe that is widely used for measuring membrane potentials of many biological systems.
  • Slow-response probe
  • Large optical response
  • Multiple applications
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty