Would you like to log in again?
Cell Navigator® Live Cell RNA Imaging Kit *Green Fluorescence*, AAT Bioquest
Supplier: AAT Bioquest
22630
76483-642EA 361.89 USD InStock
76483-642
Cell Navigator® Live Cell RNA Imaging Kit *Green Fluorescence*, AAT Bioquest
Assays Cellular Assays
Detecting and imaging RNA molecules in living cells is extremely important for a wide variety of molecular biology procedures including physical transportation, interpretation of genetic information, regulation of gene expression and some essential bio-catalytic roles.
  • RNA-selective staining
  • Improved signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty