Would you like to log in again?
Cell Navigator® Cell Plasma Membrane Staining Kit *Red Fluorescence*, AAT Bioquest
Supplier: AAT Bioquest
22681
76483-708EA 240.68 USD InStock
76483-708
Cell Navigator® Cell Plasma Membrane Staining Kit *Red Fluorescence*, AAT Bioquest
Assays Cellular Assays
The Cell Navigator® Plasma Membrane Stain Kit provides a fast and uniform labeling of the plasma membrane without the cell-type differences exhibited by lectins.
  • Signal persists through fixation
  • Stable labeling regardless of cell type
  • Multiple applications
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty