Would you like to log in again?
ReadiUse™ Stayright™ Purple *HRP Chromogen Premixed with Hydrogen Peroxide*, AAT Bioquest
Supplier: AAT Bioquest
45900 45901
76484-800EA 240.68 USD InStock
76484-800 76484-802
ReadiUse™ Stayright™ Purple *HRP Chromogen Premixed with Hydrogen Peroxide*, AAT Bioquest
Stains Microbiology Stains
3,3'-Diaminobenzidine (DAB) has been used as the most commonly used IHC chromogen because it is inexpensive and sensitive for routine applications.
  • Ready to use
  • Non-mutagenic
  • Minimal cytotoxicity
  • No mixing steps required
  • Stain persists through dehydration and coverslipping
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty